cis-4,7,10,13,16,19-Docosahexaenoic acid 98%

Synonyms: Docosa-4Z,7Z,10Z,13Z,16Z,19Z-Hexaenoic AcidDHA

AC42764-0010
103514-986EA 412 USD
103514-986
cis-4,7,10,13,16,19-Docosahexaenoic acid 98%
cis-4,7,10,13,16,19-Docosahexaenoic acid
Formula: C₂₂H₃₂O₂
MW: 328.50 g/mol
Melting Pt: –44 °C
Density: 0.95 g/cm³
Flash Pt: 62 °C
Storage Temperature: Freezer
MDL Number: MFCD00065722
CAS Number: 6217-54-5

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Viscous liquid
Infrared spectrum Conforms
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR