cis-1,4-Diacetoxy-2-butene 95%

Warning

Synonyms: cis-2-Butene-1,4-diol Diacetate(Z)-But-2-ene-1,4-diyl diacetate

AC43126-0250
103515-358EA 80.8 USD
103515-358
cis-1,4-Diacetoxy-2-butene 95%
cis-1,4-Diacetoxy-2-butene
Formula: C₈H₁₂O₄
MW: 172.18 g/mol
Boiling Pt: 222…223 °C
Density: 1.08 g/cm³
Flash Pt: >113 °C
Storage Temperature: Ambient
MDL Number: MFCD00059339
CAS Number: 25260-60-0

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
GC ≥94.0 %
GC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR