Cholesterol benzoate 98%

Synonyms: Cholesteryl benzoate

AC11020-0250 AC11020-1000
103508-590EA 172 USD
103508-590 103508-588
Cholesterol benzoate 98%
Cholesterol benzoate
Formula: C₃₄H₅₀O₂
MW: 490.77 g/mol
Melting Pt: 149…150 °C
Storage Temperature: Ambient
MDL Number: MFCD00003635
CAS Number: 604-32-0

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %
Titration RCOCl ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR