Chlorosulfuric acid 97%

Danger

Synonyms: Chlorosulphonic acidChlorosulfonic acid

AC30449-1000
103527-458EA 61.15 USD
103527-458
Chlorosulfuric acid 97%
Chlorosulphuric acid
Formula: HClO₃S
MW: 116.52 g/mol
Boiling Pt: 151…152 °C (760 mmHg)
Melting Pt: –80 °C
Density: 1.75 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00011523
CAS Number: 7790-94-5
EINECS: 232-234-6
UN: 1754
ADR: 8,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR