Chloroacetyl chloride 98%

Danger

AC14729-1000 AC14729-2500 AC14729-0025 AC14729-0010
103525-644EA 42.8 USD
103525-644 103525-640 103525-642 103525-638
Chloroacetyl chloride 98%
Chloroacetyl chloride
Formula: C₂H₂Cl₂O
MW: 112.94 g/mol
Boiling Pt: 105 °C (760 mmHg)
Melting Pt: –22 °C
Density: 1.42 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00000725
CAS Number: 79-04-9
EINECS: 201-171-6
UN: 1752
ADR: 6.1,

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR