Cesium carbonate 99.99% (trace metals basis)

Danger

AC27802-0100 AC27802-0500
103517-776EA 109 USD
103517-776 103517-774
Cesium carbonate 99.99% (trace metals basis)
Cesium carbonate
Formula: Cs₂CO₃
MW: 325.82 g/mol
Melting Pt: >400 °C
Storage Temperature: Ambient
MDL Number: MFCD00010957
CAS Number: 534-17-8
EINECS: 208-591-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR