Cefotetan 95+%

Danger

AC45949-0010 AC45949-2500
103518-632EA 74.7 USD
103518-632 103518-630
Cefotetan 95+%
Cefotetan
Formula: C₁₇H₁₇N₇O₈S₄
Storage Temperature: Refrigerator
MDL Number: MFCD00864983
CAS Number: 69712-56-7

Order Now

Specification Test Results

Appearance White to light yellow Crystalline powder
Infrared spectrum Conforms
Water ≤5 %
HPLC ≥95.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR