Cefotaxime sodium salt 95%

Danger

AC45495-0010 AC45495-0250 AC45495-0050
103467-034EA 211 USD
103467-034 103467-032 103467-072
Cefotaxime sodium salt 95%
Cefotaxime sodium salt
Formula: C₁₆H₁₇N₅O₇S₂
Melting Pt: 201…210 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00079073
CAS Number: 64485-93-4

Order Now

Specification Test Results

Appearance White to light yellow Powder
Infrared spectrum Conforms
Loss on drying ≤3.0 % (105 °C, 8 hrs)
HPLC ≥94 %
pH 4.5 to 6.5
Specific optical rotation +58° to +65° (20 °C, 589 nm) (c=1, H2O)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR