Calcium stearate (6.4 - 7.4% Ca), powder

Synonyms: Stearic acid calcium salt

AC44972-0010 AC44972-0050
103517-166EA 201 USD
103517-166 103517-164
Calcium stearate (6.4 - 7.4% Ca), powder
Calcium stearate
Formula: C₃₆H₇₀CaO₄
MW: 607.03 g/mol
Melting Pt: 145…160 °C
Storage Temperature: Ambient
MDL Number: MFCD00036390
CAS Number: 1592-23-0
https://us.vwr.com/stibo/web/std.lang.all/25/69/2302569.jpg

Order Now

Specification Test Results

Appearance White to off-white Powder
Titration Complexometric 6.4 to 7.5 % (Ca)
Loss on drying ≤6 % (105 °C, 3 hrs)
Ash 9 % to 10.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR