Calcium bis(dihydrogenorthophosphate) ≥90%, pure

Supplier: ACROS ORGANICS MS

Danger

Synonyms: Calcium phosphate, monobasic

AC37861-0010
103463-484EA 45.97 USD
103463-484
Calcium bis(dihydrogenorthophosphate) ≥90%, pure
Calcium bis(dihydrogenorthophosphate)
Formula: H₄CaO₈P₂
Storage Temperature: Ambient
MDL Number: MFCD00010898
CAS Number: 7758-23-8

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥88 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR