tri-Calcium phosphate 96%, pure

Synonyms: Calcium phosphate tribasicpenta-Calcium hydroxide triphosphateTricalcium orthophosphateCalcium phosphate

AC44639-0050
103464-136EA 395 USD
103464-136
tri-Calcium phosphate 96%, pure
tri-Calcium phosphate
Formula: Ca₃(PO₄)₂
MW: 310.18 g/mol
Melting Pt: 1670 °C
Storage Temperature: Ambient
MDL Number: MFCD00015984
CAS Number: 7758-87-4

Order Now

Specification Test Results

Appearance White Powder
Titration Complexometric ≥95 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR