Cadmium oxide 99%, pure

Danger

Synonyms: Cadmium (II) oxide

AC22379-0050
103526-994EA 277 USD
103526-994
Cadmium oxide 99%, pure
Cadmium oxide
Formula: CdO
MW: 128.41 g/mol
Boiling Pt: 1385 °C (760 mmHg)
Storage Temperature: Ambient
MDL Number: MFCD00010921
CAS Number: 1306-19-0
EINECS: 215-146-2
UN: 2570
ADR: 6.1,I
Merck Index: 12,01659

Order Now

Specification Test Results

Appearance Red-brown to brown Powder
Titration Complexometric ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR