Bis(pentafluorophenyl)phenylphosphine 90% tech.

Warning

Synonyms: Bis(perfluorophenyl)(phenyl)phosphineSpectraSynQ 443

AC31317-1000
103512-302EA 76.43 USD
103512-302
Bis(pentafluorophenyl)phenylphosphine 90% tech.
Bis(pentafluorophenyl)phenylphosphine
Formula: C₁₈H₅F₁₀P
MW: 442.2 g/mol
Boiling Pt: 105 °C (.2 mmHg)
Melting Pt: 56…62 °C
Flash Pt: >110 °C
Storage Temperature: Ambient
MDL Number: MFCD00000291
CAS Number: 5074-71-5
EINECS: 225-780-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR