Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal™

Danger

Synonyms: 2-Methoxyethyl etherDiethylene glycol dimethyl ether1-Methoxy-2-(2-methoxyethoxy)ethaneDiglymeDimethyldigol

AC44310-1000 AC44310-0010
103528-888EA 283 USD
103528-888 103528-890
Bis(2-methoxyethyl)ether, extra dry over Molecular Sieve 99+%, AcroSeal™
Bis(2-methoxyethyl) ether
Formula: C₆H₁₄O₃
MW: 134.18 g/mol
Boiling Pt: 162 °C (760 mmHg)
Melting Pt: –64 °C
Density: 0.937 g/cm³
Flash Pt: 51 °C
Storage Temperature: Ambient
MDL Number: MFCD00008503
CAS Number: 111-96-6
UN: 3271
ADR: 3,III

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥99.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR