Estradiol 98%

Danger

Synonyms: beta-Estradiol17β-Estradiol

AC43632-0050 AC43632-0250
103513-190EA 798 USD
103513-190 103515-796
Estradiol 98%
Estradiol
Formula: C₁₈H₂₄O₂
MW: 272.39 g/mol
Melting Pt: 176…180 °C
Storage Temperature: Ambient
MDL Number: MFCD00003693
CAS Number: 50-28-2

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 173 to 179 °C
Water ≤3.5 %
HPLC ≥97.5 %
Specific optical rotation +76° to +83° (20 °C, 589 nm) (c=1, dioxane)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR