Dibenzyl sulfoxide ≥98%

Synonyms: Benzyl sulfoxideDibenzyl SulfoxideBenzyl sulphoxideDibenzyl sulphoxide

AC10617-5000
103508-456EA 93.4 USD
103508-456
Dibenzyl sulfoxide ≥98%
Dibenzyl sulphoxide
Formula: C₁₄H₁₄OS
MW: 230.33 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00004782
CAS Number: 621-08-9

Order Now

Specification Test Results

Appearance Off-white to light yellow Crystalline powder
Infrared spectrum Conforms
Melting point ≥130 °C
HPLC ≥98.0 %
Molar absorption coefficient ≥42000 l/(mol.cm) (at Lambda max)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR