Atenolol 98%

Synonyms: 2-[4-(2-Hydroxy-3-isopropylaminopropoxy)phenyl]acetamide

AC44955-0010 AC44955-0050 AC44955-0250
103517-140EA 56.6 USD
103517-140 103517-144 103517-142
Atenolol 98%
Atenolol
Formula: C₁₄H₂₂N₂O₃
MW: 266.34 g/mol
Melting Pt: 146…148 °C
Storage Temperature: Ambient
MDL Number: MFCD00057645
CAS Number: 29122-68-7

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Melting point 151 to 155 °C
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR