Anisole, extra dry 99%, AcroSeal™

Warning

Synonyms: MethoxybenzeneMethyl phenyl ether

AC42925-0010 AC42925-1000
103524-354EA 96.7 USD
103524-354 103524-352
Anisole, extra dry 99%, AcroSeal™
Anisole
Formula: C₇H₈O
MW: 108.14 g/mol
Boiling Pt: 154 °C (760 mmHg)
Melting Pt: –37 °C
Density: 0.99 g/cm³
Flash Pt: 45 °C
Storage Temperature: Ambient
MDL Number: MFCD00008354
CAS Number: 100-66-3
UN: 2222
ADR: 3,III
https://us.vwr.com/stibo/web/std.lang.all/96/93/759693.jpg

Order Now

Specification Test Results

Appearance Clear colorless Liquid
Infrared spectrum Conforms
Water ≤0.005 %
GC ≥98.5 %
Residue after evaporation ≤0.005 %
UV at 200 nm A: ≤0.75
UV 1 cm cell vs HPLC-grade water
Non-volatile matter ≤0.0003 %
Free acid (CH3COOH) ≤0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR