Ammonium trifluoromethanesulfonate 99%

Warning

Synonyms: Ammonium triflateMethanesulfonic acid,1,1,1-trifluoro-, ammonium salt (1:1)Methanesulphonic acid,1,1,1-trifluoro-, ammonium salt (1:1)

AC38873-0500
103464-062EA 225 USD
103464-062
Ammonium trifluoromethanesulfonate 99%
Ammonium trifluoromethanesulfonate
Formula: CH₄F₃NO₃S
MW: 167.102 g/mol
Melting Pt: 225 °C
Storage Temperature: Ambient
MDL Number: MFCD00075332
CAS Number: 38542-94-8

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
Elemental Analysis ≥98.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR