Ammonium metavanadate 99.996% (trace metals basis)

Danger

Synonyms: Ammonium vanadium oxideAmmonium monovanadateAmmonium vanadium trioxide

AC19440-0500
103526-460EA 474 USD
103526-460
Ammonium metavanadate 99.996% (trace metals basis)
Ammonium metavanadate
Formula: NH₄VO₃
MW: 116.98 g/mol
Melting Pt: 200 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00011430
CAS Number: 7803-55-6
UN: 2859
ADR: 6.1,II

Order Now

Specification Test Results

Appearance White to yellow Crystalline powder
Total trace metal impurities ≤40 ppm
Vanadium (V) 42.4 to 44.6 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR