Amberlyst® A-26 (OH) ion-exchange resin

Danger

AC20218-0010 AC20218-1000
103509-640EA 134 USD
103509-640 103509-642
Amberlyst® A-26 (OH) ion-exchange resin
Amberlyst® A-26 (OH)
Density: 0.672 g/cm³
Storage Temperature: Ambient
MDL Number: MFCD00145579
CAS Number: 39339-85-0

Order Now

Specification Test Results

Appearance Tan Beads
Harmonic mean size 0.560 to 0.700
Uniformity coefficient ≤1.45
Moisture holding capacity 65 to 75 % (OH)
Total volume capacity ≥0.80 eq/l (OH) as is

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR