Amberlite® IRN-78, Ion exchange resin cationic type (OH-) ion-exchange resin

Danger

AC42929-0025 AC42929-5000
103467-298EA 363.46 USD
103467-298 103467-302
Amberlite® IRN-78, Ion exchange resin cationic type (OH-) ion-exchange resin
AMBERLITE® IRN-78, Ion exchange resin cationic type (OH-)
Storage Temperature: Ambient
MDL Number: MFCD00145822
CAS Number: 11128-95-3

Order Now

Specification Test Results

Appearance Yellow to amber Spherical beads
Harmonic mean size 0.580 to 0.680 mm
Uniformity coefficient ≤1.20
Moisture holding capacity 54.0 to 60.0 % (OH)
Total volume capacity ≥1.10 eq/l (OH) as is

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR