α-Ketoglutaric acid monosodium salt 98%

Warning

AC43935-1000 AC43935-0050 AC43935-0250
103518-902EA 58.5 USD
103518-902 103518-904 103518-906
α-Ketoglutaric acid monosodium salt 98%
α-Ketoglutaric acid monosodium salt

Order Now

Specification Test Results

Appearance White Powder
Infrared spectrum Conforms
Loss on drying ≤1.0 %
UV at 430 nm A: ≤0.200
pH 3.6 to 4.0 (0.5 % in water)
Titration with HClO4 ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR