Allylamine 98%

Danger

AC22073-0500 AC22073-2500
103526-882EA 103 USD
103526-882 103526-880
Allylamine 98%
Allylamine
Formula: C₃H₇N
MW: 57.1 g/mol
Boiling Pt: 53 °C (760 mmHg)
Melting Pt: –88 °C
Density: 0.763 g/cm³
Flash Pt: –28 °C
Storage Temperature: Ambient
MDL Number: MFCD00008199
CAS Number: 107-11-9
EINECS: 203-463-9
UN: 2334
ADR: 6.1,

Order Now

Specification Test Results

Appearance Clear colorless to light yellow Liquid
Infrared spectrum Conforms
Refractive index 1.4200 to 1.4230 (20 °C, 589 nm)
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR