Alizarine yellow R (sodium salt) 96%, pure

Warning

AC40049-0250
103518-824EA 33.5 USD
103518-824
Alizarine yellow R (sodium salt) 96%, pure
Alizarine Yellow R (sodium salt)
Formula: C₁₃H₈N₃NaO₅
Melting Pt: 253…254 °C
Storage Temperature: Ambient
MDL Number: MFCD00067121
CAS Number: 1718-34-9
EINECS: 217-002-4

Order Now

Specification Test Results

Appearance Orange to brown Crystalline powder
Infrared spectrum Conforms
Loss on drying ≤5 % (105 °C)
UV spectrophotometry ≥95 %
Visual transition interval from pH 10.2 (yellow) to pH 12.0 (red)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR