Acrylonitrile 99+%

Danger

AC14963-0025 AC14963-1000 AC14963-0100 AC14963-0010
103525-742EA 41.78 USD
103525-742 103525-740 103525-736 103525-738
Acrylonitrile 99+%
Acrylonitrile
Formula: C₃H₃N
MW: 53.06 g/mol
Boiling Pt: 77.3 °C (760 mmHg)
Melting Pt: –83.5 °C
Density: 0.8 g/cm³
Flash Pt: –0.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00001927
CAS Number: 107-13-1
EINECS: 203-466-5
UN: 1093
ADR: 3,I

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR