Ammonium acetate-[D₇] (99% D)
|
Warning
|
Synonyms:
(2H3)Acetic acid (2H4)ammonium salt
, (2H4)Ammonium (2H3)acetate
, Acetic acid ammonium salt-D7
, (2H7)Acetic acid ammonium salt
, (2H7)Ammonium acetate
, Ammonium acetate-[²H₇]
AC42586-0010
AC42586-0050
103518-016EA
222.69
USD
103518-016
103518-014
Ammonium acetate-[D₇] (99% D)
Ammonium acetate-[D7]
Formula:
D₃CCOOND₄ MW: 84.03 g/mol Melting Pt: 110…113 °C Storage Temperature: Ambient |
MDL Number:
MFCD00190378 CAS Number: 194787-05-8 |
Specification Test Results
Appearance | White Crystalline powder |
D-Enrichment | ≥97.5 % |
Melting point | 95.0 to 100.0 °C |
Elemental Analysis | ≥96.0 % (ex C) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...