1-[(9H-Fluoren-9-ylmethoxy)carbonyloxy]benzotriazole 98%

Warning

Synonyms: 1-(Fmoc-oxy)benzotriazoleFmoc-OBt(9H-Fluoren-9-yl)methyl 1H-benzo[d][1,2,3]triazol-1-yl carbonateCarbonic acid 1-Benzotriazolyl 9-Fluorenylmethyl Ester9-Fluorenylmethyl 1-Benzotriazolyl Carbonate1-(Fmoc-oxy)-1H-benzotriazole

AC44151-0050
103466-848EA 166.4 USD
103466-848
1-[(9H-Fluoren-9-ylmethoxy)carbonyloxy]benzotriazole 98%
1-[(9H-Fluoren-9-ylmethoxy)carbonyloxy]benzotriazole
Formula: C₂₁H₁₅N₃O₃
MW: 357.37 g/mol
Melting Pt: 165…167 °C
Storage Temperature: Ambient
MDL Number: MFCD00074939
CAS Number: 82911-71-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Elemental Analysis 68.82 to 72.34 % (C)
Elemental Analysis 11.46 to 12.05 % (N)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR