8-Bromoimidazo[1,2-a]pyridine 95%

Warning

AC44661-0050
103516-654EA 424.9 USD
103516-654
8-Bromoimidazo[1,2-a]pyridine 95%
8-Bromoimidazo[1,2-a]pyridine
Formula: C₇H₅BrN₂
MW: 197.03 g/mol
Melting Pt: 72…75 °C
Storage Temperature: Refrigerator
MDL Number: MFCD08706060
CAS Number: 850349-02-9

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Melting point 70 to 75 °C
GC ≥94.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR