8-Anilino-1-naphthalenesulfonic acid 98%

Warning

Synonyms: 8-(Phenylamino)naphthalene-1-sulphonic acid8-Anilinonaphthalene-1-sulfonic acid8-Anilinonaphthalene-1-sulphonic acid8-(Phenylamino)naphthalene-1-sulfonic acid

AC40121-1000 AC40121-0050 AC40121-0250
103514-656EA 51.2 USD
103514-656 103514-658 103514-660
8-Anilino-1-naphthalenesulfonic acid 98%
8-Anilino-1-naphthalenesulphonic acid
Formula: C₁₆H₁₃NO₃S
MW: 299.35 g/mol
Melting Pt: 183…183.5 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00003998
CAS Number: 82-76-8

Order Now

Specification Test Results

Appearance Dark green to brown to black Powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR