7-Methoxy-2-naphthol 97%

Warning

Synonyms: 7-Methoxynaphthalen-2-ol

AC42887-0250 AC42887-0050
103515-050EA 243 USD
103515-050 103515-048
7-Methoxy-2-naphthol 97%
7-Methoxy-2-naphthol

Order Now

Specification Test Results

Appearance White to brown Crystalline powder and/or chunks
Infrared spectrum Conforms
Melting point 116 to 119 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR