7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%

Warning

AC45046-0050 AC45046-0010
103518-254EA 166 USD
103518-254 103518-256
7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride 97%
7-Fluoro-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formula: C₉H₁₀FN·HCl
MW: 187.65 g/mol
Melting Pt: 177…179 °C
Storage Temperature: Ambient
MDL Number: MFCD08461074
CAS Number: 799274-06-9

Order Now

Specification Test Results

Appearance White to light brown Powder
Infrared spectrum Conforms
Titration Argentometric ≥96.0 to ≤104.0 %
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR