6-Methylnicotinamide 97%

Warning

Synonyms: 6-Methylpyridine-3-carboxamide

AC45368-0050 AC45368-0010
103518-380EA 216.4 USD
103518-380 103518-378
6-Methylnicotinamide 97%
6-Methylnicotinamide
Formula: C₇H₈N₂O
MW: 136.15 g/mol
Melting Pt: 197…199 °C
Storage Temperature: Ambient
MDL Number: MFCD00006342
CAS Number: 6960-22-1

Order Now

Specification Test Results

Appearance White to yellow Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR