6-Fluoropicolinic acid 97%
|
Warning
|
Synonyms:
2-Fluoro-6-pyridinecarboxylic acid
, 6-Fluoropyridine-2-carboxylic acid
AC43684-0050
103465-810EA
913
USD
103465-810
6-Fluoropicolinic acid 97%
6-Fluoropicolinic acid
Formula:
C₆H₄FNO₂ MW: 141.1 g/mol Melting Pt: 138.4…141.1 °C Storage Temperature: Ambient |
MDL Number:
MFCD02181193 CAS Number: 402-69-7 |
Specification Test Results
Appearance | White to off-white Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 135 to 142 °C |
Water | ≤1.0 % |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...