2-Fluoro-5-pyridineboronic acid 97%

Warning

Synonyms: 2-Fluoro-5-pyridylboronic acid2-Fluoropyridine-5-boronic acid6-Fluoro-3-pyridinylboronic acid2-Fluoro-5-pyridinylboronic acid6-Fluoropyridine-3-boronic acid

AC39716-0050 AC39716-0010
103463-758EA 54.7 USD
103463-758 103463-760
2-Fluoro-5-pyridineboronic acid 97%
2-Fluoro-5-pyridineboronic acid
Formula: C₅H₅BFNO₂
MW: 140.91 g/mol
Melting Pt: 172…176 °C
Storage Temperature: Ambient
MDL Number: MFCD03411559
CAS Number: 351019-18-6

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
Titration with NaOH ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR