6-Ethoxy-2-naphthaleneboronic acid 97%

Warning

Synonyms: (6-Ethoxynaphthalen-2-yl)boronic acid

AC44023-0050 AC44023-0010
103463-756EA 139 USD
103463-756 103463-754
6-Ethoxy-2-naphthaleneboronic acid 97%
6-Ethoxy-2-naphthaleneboronic acid
Formula: C₁₂H₁₃BO₃
MW: 216.04 g/mol
Melting Pt: 238.5…243.3 °C
Storage Temperature: Ambient
MDL Number: MFCD03427009
CAS Number: 352525-98-5

Order Now

Specification Test Results

Appearance White to off-white Powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR