6-Chloro-2-hydroxypyridine 98%

Warning

Synonyms: 6-Chloropyridin-2(1H)-one6-Chloro-2-pyridinol2-Chloro-6-pyridinol2-Chloro-6-hydroxypyridine

AC15966-0250
103465-770EA 54.3 USD
103465-770
6-Chloro-2-hydroxypyridine 98%
6-Chloro-2-hydroxypyridine
Formula: C₅H₄ClNO
MW: 129.55 g/mol
Melting Pt: 129.2…129.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00006278
CAS Number: 16879-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR