6-Bromoisoquinoline 97%

Warning

AC42946-0010 AC42946-0050
103515-082EA 272 USD
103515-082 103515-080
6-Bromoisoquinoline 97%
6-Bromoisoquinoline
Formula: C₉H₆BrN
Melting Pt: 42.6…43.6 °C
Storage Temperature: Ambient
MDL Number: MFCD04973299
CAS Number: 34784-05-9

Order Now

Specification Test Results

Appearance Orange to light brown Low melting solid
Infrared spectrum Conforms
Melting point 40 to 45 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR