6-Bromo-1-methyl-1H-indazole 97%

Warning

AC44460-0010 AC44460-0050
103513-702EA 1080.2 USD
103513-702 103513-700
6-Bromo-1-methyl-1H-indazole 97%
6-Bromo-1-methyl-1H-indazole
Formula: C₈H₇BrN₂
MW: 211.0600 g/mol
Melting Pt: 48…50 °C
Storage Temperature: Ambient
MDL Number: MFCD09878569
CAS Number: 590417-94-0

Order Now

Specification Test Results

Appearance White to yellow Solid
Infrared spectrum Conforms
Melting point 48 to 53 °C
GC ≥96 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR