6-Bromo-1H-indazole 97%

Warning

AC44459-0010
103465-726EA 38 USD
103465-726
6-Bromo-1H-indazole 97%
6-Bromo-1H-indazole
Formula: C₇H₅BrN₂
MW: 197.0300 g/mol
Melting Pt: 181…185 °C
Storage Temperature: Ambient
MDL Number: MFCD03265457
CAS Number: 79762-54-2

Order Now

Specification Test Results

Appearance Off-white to brown Solid
Infrared spectrum Conforms
Melting point 180 to 186 °C
HPLC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR