6,7-Dimethoxy-1-tetralone 97%

Warning

Synonyms: 6,7-Dimethoxy-3,4-dihydronaphthalen-1(2H)-one

AC44020-0050 AC44020-0010
103465-668EA 223 USD
103465-668 103513-326
6,7-Dimethoxy-1-tetralone 97%
6,7-Dimethoxy-1-tetralone
Formula: C₁₂H₁₄O₃
MW: 206.24 g/mol
Melting Pt: 95…98 °C
Storage Temperature: Ambient
MDL Number: MFCD00134100
CAS Number: 13575-75-2

Order Now

Specification Test Results

Appearance White to off-white to beige Crystalline powder or crystals
Infrared spectrum Conforms
Melting point 95 to 100 °C
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR