6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%

Warning

AC29298-0050
103465-664EA 43.39 USD
103465-664
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride 98%
6,7-Dimethoxy-1,2,3,4-tetrahydroisoquinoline hydrochloride
Formula: C₁₁H₁₆ClNO₂
MW: 229.71 g/mol
Melting Pt: 252…254 °C
Storage Temperature: Ambient
MDL Number: MFCD00012744
CAS Number: 2328-12-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR