5-Pyrimidineacetic acid 95%

Warning

AC45428-0010 AC45428-2500
103465-644EA 169.2 USD
103465-644 103465-642
5-Pyrimidineacetic acid 95%
5-Pyrimidineacetic acid
Formula: C₆H₆N₂O₂
MW: 138.1240 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09608122
CAS Number: 5267-07-2

Order Now

Specification Test Results

Appearance Yellow to brown Powder or solid
Infrared spectrum Conforms
HPLC ≥94.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR