5-Fluorosalicylaldehyde 97%

Warning

Synonyms: 5-Fluoro-2-hydroxybenzaldehyde

AC39757-0010 AC39757-0050
103465-584EA 103 USD
103465-584 103465-586
5-Fluorosalicylaldehyde 97%
5-Fluorosalicylaldehyde
Formula: C₇H₅FO₂
MW: 140.11 g/mol
Melting Pt: 82…85 °C
Storage Temperature: Ambient
MDL Number: MFCD01090997
CAS Number: 347-54-6

Order Now

Specification Test Results

Appearance White to light brown Crystalline powder or solid
Infrared spectrum Conforms
Melting point 81 to 86 °C
GC ≥96.0 %
NMR (1H-NMR) Consistent with structure

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR