5-Fluoro-2-methoxyphenylboronic acid 98%

Warning

Synonyms: 5-Fluoro-2-methoxybenzeneboronic acid

AC39389-0010 AC39389-0050
103465-552EA 20 USD
103465-552 103465-554
5-Fluoro-2-methoxyphenylboronic acid 98%
5-Fluoro-2-methoxyphenylboronic acid
Formula: C₇H₈BFO₃
MW: 169.95 g/mol
Melting Pt: 156…158 °C
Storage Temperature: Ambient
MDL Number: MFCD01863526
CAS Number: 179897-94-0

Order Now

Specification Test Results

Appearance White to off-white Powder or crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR