5-Cyano-2-fluorophenylboronic acid 97%

Warning

Synonyms: 5-Cyano-2-fluorobenzeneboronic acid

AC43023-0010 AC43023-0050
103465-538EA 229.5 USD
103465-538 103465-540
5-Cyano-2-fluorophenylboronic acid 97%
5-Cyano-2-fluorophenylboronic acid
Formula: C₇H₅BFNO₂
MW: 164.9300 g/mol
Melting Pt: 350…353 °C
Storage Temperature: Ambient
MDL Number: MFCD03095147
CAS Number: 468718-30-1

Order Now

Specification Test Results

Appearance White Crystalline powder
Infrared spectrum Conforms
HPLC ≥96.0 %
GC ≥99.50 %
Color scale ≤50 APHA
Chloride (Cl) ≤1 ppm
Aluminium (Al) ≤0.5 ppm
Barium (Ba) ≤0.1 ppm
Calcium (Ca) ≤0.5 ppm
Iron (Fe) ≤0.1 ppm
Lead (Pb) ≤0.1 ppm
Magnesium (Mg) ≤0.1 ppm
Nickel (Ni) ≤0.02 ppm
Zinc (Zn) ≤0.1 ppm
Free amine ≤0.01 % (as CH3NH2)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR