5-Chloro-3-methylbenzo[b]thiophene 97%

Warning

AC45099-0050 AC45099-0010 AC45099-0250
103465-518EA 36.1 USD
103465-518 103465-520 103465-522
5-Chloro-3-methylbenzo[b]thiophene 97%
5-Chloro-3-methylbenzo[b]thiophene
Formula: C₉H₇ClS
Boiling Pt: 87 °C (.5 mmHg)
Melting Pt: 31…32 °C
Storage Temperature: Ambient
MDL Number: MFCD00052506
CAS Number: 19404-18-3

Order Now

Specification Test Results

Appearance White to yellow Low melting solid
Infrared spectrum Conforms
Melting point 30 to 36 °C
GC ≥96.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR