5-Chloro-2-hydroxypyridine 97%

Warning

Synonyms: 3-Chloro-6-hydroxypyridine3-Chloro-6-pyridinol5-Chloro-2-pyridinol

AC15939-0100
103465-516EA 29.1 USD
103465-516
5-Chloro-2-hydroxypyridine 97%
5-Chloro-2-hydroxypyridine
Formula: C₅H₄ClNO
MW: 129.55 g/mol
Melting Pt: 164.4…165.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00006275
CAS Number: 4214-79-3
EINECS: 224-146-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR