5-Bromooxindole 97%
|
Warning
|
Synonyms:
5-Bromo-1,3-dihydro-2H-indol-2-one
, 5-Bromoindolin-2-one
, 5-Bromo-2-oxindole
AC39853-0010
103465-486EA
71.7
USD
103465-486
5-Bromooxindole 97%
5-Bromooxindole
Formula:
C₈H₆BrNO MW: 212.05 g/mol Melting Pt: 219…221 °C Storage Temperature: Ambient |
MDL Number:
MFCD00456998 CAS Number: 20870-78-4 |
Specification Test Results
Appearance | Off-white to tan Powder or crystals |
Infrared spectrum | Conforms |
Melting point | 220 to 224 °C |
HPLC | ≥96.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...