5-Bromoindole-2-carboxylic acid 98%

Warning

Synonyms: 5-Bromo-1H-indole-2-carboxylic acid

AC39865-2500 AC39865-0050 AC39865-0010
103465-468EA 46.3 USD
103465-468 103465-464 103465-466
5-Bromoindole-2-carboxylic acid 98%
5-Bromoindole-2-carboxylic acid
Formula: C₉H₆BrNO₂
MW: 240.06 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00022705
CAS Number: 7254-19-5

Order Now

Specification Test Results

Appearance White to beige Crystalline powder
Infrared spectrum Conforms
HPLC ≥97.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR